siRNA Details
| Gene Symbol: Rab33a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011228.2 | NP_035358.1 | 1, 2 | 567 | ||
| GenBank mRNA | AB232628.1 | - | 1, 2 | 251 | ||
| BC104390.1 | - | 1, 2 | 315 | 40050293 | ||
| BC104391.1 | - | 1, 2 | 315 | 40050296 | ||
| D83279.1 | - | 1, 2 | 382 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001108257.

Rab33a
Genomic Map