siRNA Details
| Gene Symbol: Rad17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044371.2 | NP_001037836.1 | 3 | 385 | ||
| NM_001283011.1 | NP_001269940.1 | 3 | 385 | |||
| NM_011233.3 | NP_035363.2 | 3 | 513 | |||
| GenBank mRNA | AF085737.1 | - | 3 | 436 | ||
| AJ011923.1 | - | 2 | 286 | |||
| AK134277.1 | - | 3 | 340 | |||
| AK162016.1 | - | 3 | 372 | |||
| AK167358.1 | - | 3 | 328 | |||
| BC023939.1 | - | 3 | 390 | |||
| BC066855.1 | - | 3 | 503 | 30473984 | ||
| CF916039.1 | - | 3 | 443 | |||

Rad17
Genomic Map