siRNA Details
| Gene Symbol: Rasgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011245.2 | NP_035375.1 | 3, 4 | 765 | ||
| GenBank mRNA | AK079638.1 | - | 3, 4 | 648 | ||
| AK140380.1 | - | 3, 4 | 632 | |||
| BC138682.1 | - | 3, 4 | 601 | 8861703 | ||
| BC138683.1 | - | 3, 4 | 601 | 8861704 | ||
| L20899.1 | - | 3, 4 | 767 | |||

Rasgrf1
Genomic Map