siRNA Details
| Gene Symbol: Rasgrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011246.2 | NP_035376.1 | 4 | 509 | ||
| GenBank mRNA | AF106070.1 | - | 4 | 353 | ||
| AK013548.1 | - | 4 | 555 | |||
| AK134005.1 | - | 4 | 523 | |||
| AK141524.1 | - | 4 | 634 | |||
| BC057120.1 | - | 4 | 509 | 6831137 | ||
| BC057341.1 | - | 4 | 509 | 6831137 | ||

Rasgrp1
Genomic Map