siRNA Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011161.5 | NP_035291.4 | 11, 12 | 1115 | ||
| GenBank mRNA | AF135185.1 | - | 11, 12 | 1095 | ||
| AK144846.1 | - | 11, 12 | 1088 | |||
| AK161190.1 | - | 9, 10 | 1290 | |||
| AK161562.1 | - | 11, 12 | 1067 | |||
| BC057211.1 | - | 11, 12 | 1051 | |||
| BC064737.1 | - | 11, 12 | 1066 | 30112690 | ||
| BC092526.1 | - | 11, 12 | 1115 | 6593321 | ||

Mapk11
Genomic Map