siRNA Details
Gene Symbol: NKX6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152568.3 | NP_689781.1 | 2 | 749 | ||
XM_017013143.1 | XP_016868632.1 | 3 | 1461 | |||
GenBank mRNA | AK057898.1 | - | 3 | 706 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NKX6-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152568.3 | NP_689781.1 | 2 | 749 | ||
XM_017013143.1 | XP_016868632.1 | 3 | 1461 | |||
GenBank mRNA | AK057898.1 | - | 3 | 706 |