siRNA Details
| Gene Symbol: Rbm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011251.3 | NP_035381.2 | 19 | 3314 | ||
| NM_029169.3 | NP_083445.1 | 19 | 3585 | |||
| GenBank mRNA | AJ006486.2 | - | 19 | 3190 | ||
| AK077899.1 | - | 19 | 3493 | |||
| AK145612.1 | - | 19 | 3474 | |||
| AK164432.1 | - | 14 | 1671 | |||
| AK220446.1 | - | 19 | 3460 | |||
| BC026129.1 | - | 19 | 3351 | 3982861 | ||

Rbm6
Genomic Map