siRNA Details
| Gene Symbol: Trim10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011280.2 | NP_035410.2 | 1 | 455 | ||
| GenBank mRNA | AF134811.1 | - | 1 | 437 | ||
| AF220121.1 | - | 1 | 442 | |||
| AK011082.1 | - | 1 | 455 | |||
| BC051632.1 | - | 1 | 428 | 6507331 | ||
| BY712181.1 | - | 1 | 456 | |||

Trim10
Genomic Map