siRNA Details
| Gene Symbol: Ccl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011331.2 | NP_035461.2 | 2 | 216 | ||
| GenBank mRNA | AF065934.1 | - | 2 | 182 | ||
| AF065935.1 | - | 2 | 182 | |||
| AF065936.1 | - | 2 | 182 | |||
| AF065937.1 | - | 2 | 182 | |||
| AF065938.1 | - | 2 | 182 | |||
| AK012356.1 | - | 2 | 216 | |||
| BC027520.1 | - | 2 | 171 | 1548072 | ||
| U50712.1 | - | 2 | 215 | |||
| U66670.1 | - | 2 | 192 | |||

Ccl12
Genomic Map