siRNA Details
| Gene Symbol: Sema3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | Z93947.1 | - | 1 | 712 | ||
| Z93948.1 | - | 1 | 712 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sema3e| Gene Symbol: Sema3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | Z93947.1 | - | 1 | 712 | ||
| Z93948.1 | - | 1 | 712 | |||