siRNA Details
| Gene Symbol: Frzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011356.4 | NP_035486.1 | 4 | 1179 | ||
| GenBank mRNA | AK019093.2 | - | 4 | 850 | ||
| AK029941.1 | - | 4 | 1181 | |||
| BC016884.1 | - | 4 | 1026 | 4237554 | ||
| U68058.1 | - | 4 | 635 | |||
| U88568.1 | - | 4 | 987 | |||
| U91905.1 | - | 4 | 1421 | |||
More Information
Other Species Targeted
Set Membership: |
|

Frzb
Genomic Map