siRNA Details
| Gene Symbol: Rph3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB112924.1 | - | 5 | 385 | ||
| AK139040.1 | - | 4 | 327 | |||
| BC042585.1 | - | 5 | 552 | 5364086 | ||
| BC050883.1 | - | 5 | 519 | 5698459 | ||
| D29965.1 | - | 5 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rph3a| Gene Symbol: Rph3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB112924.1 | - | 5 | 385 | ||
| AK139040.1 | - | 4 | 327 | |||
| BC042585.1 | - | 5 | 552 | 5364086 | ||
| BC050883.1 | - | 5 | 519 | 5698459 | ||
| D29965.1 | - | 5 | 362 | |||