siRNA Details
| Gene Symbol: Ccl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011329.3 | NP_035459.1 | 3 | 284 | ||
| GenBank mRNA | AF065924.1 | - | 3 | 263 | ||
| AF065925.1 | - | 3 | 263 | |||
| AF065926.1 | - | 3 | 263 | |||
| AK019040.1 | - | 3 | 284 | |||
| BC042744.1 | - | 1895 | ||||
| BC120806.1 | - | 3 | 275 | 40129729 | ||
| M17957.1 | - | 3 | 287 | |||
| M23501.1 | - | 3 | 383 | |||

Ccl1
Genomic Map