siRNA Details
| Gene Symbol: Cxcl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011339.2 | NP_035469.1 | 2, 3 | 225 | ||
| GenBank mRNA | AF082859.1 | - | 2, 3 | 188 | ||
| AK142955.1 | - | 2, 3 | 227 | |||
| BC061138.1 | - | 2, 3 | 218 | 30298837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl15| Gene Symbol: Cxcl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011339.2 | NP_035469.1 | 2, 3 | 225 | ||
| GenBank mRNA | AF082859.1 | - | 2, 3 | 188 | ||
| AK142955.1 | - | 2, 3 | 227 | |||
| BC061138.1 | - | 2, 3 | 218 | 30298837 | ||