siRNA Details
| Gene Symbol: Sema3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011349.3 | NP_035479.2 | 4, 5 | 536 | ||
| GenBank mRNA | AF036162.1 | - | 2, 3 | 194 | ||
| AF080090.1 | - | 3, 4 | 349 | |||
| AF080091.1 | - | 3, 4 | 335 | |||
| AK053632.1 | - | 4, 5 | 537 | |||
| AK140340.1 | - | 4, 5 | 988 | |||
| BC010976.1 | - | 4, 5 | 457 | 4219060 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK172610.1 | - | 929 | Chr. - - [-] on Build | ||

Sema3f
Genomic Map