siRNA Details
| Gene Symbol: Shbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011367.2 | NP_035497.1 | 4 | 498 | ||
| GenBank mRNA | AK010959.1 | - | 4 | 498 | ||
| BC120785.1 | - | 4 | 483 | 40129708 | ||
| BC120787.1 | - | 4 | 483 | 40129710 | ||
| U85644.1 | - | 4 | 461 | |||

Shbg
Genomic Map