siRNA Details
| Gene Symbol: St6galnac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011371.2 | NP_035501.2 | 9 | 1903 | ||
| GenBank mRNA | Y11274.1 | - | 9 | 1903 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St6galnac1| Gene Symbol: St6galnac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011371.2 | NP_035501.2 | 9 | 1903 | ||
| GenBank mRNA | Y11274.1 | - | 9 | 1903 | ||