siRNA Details
| Gene Symbol: St6galnac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276425.1 | NP_001263354.1 | 6 | 3167 | ||
| NM_011373.3 | NP_035503.1 | 5 | 3520 | |||
| GenBank mRNA | AJ007310.1 | - | 6 | 3163 | ||
| AK161559.1 | - | 6 | 3152 | |||
| BC086784.1 | - | 3108 | 6844232 | |||

St6galnac4
Genomic Map