siRNA Details
| Gene Symbol: St8sia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011374.2 | NP_035504.2 | 5 | 1667 | ||
| GenBank mRNA | AK042207.1 | - | 5 | 1523 | ||
| AK046188.1 | - | 5 | 1607 | |||
| AK052444.1 | - | 5 | 1318 | |||
| BC024821.1 | - | 5 | 1667 | 5357151 | ||
| L38677.1 | - | 5 | 1291 | |||
| X84235.1 | - | 5 | 1014 | |||

St8sia1
Genomic Map