siRNA Details
| Gene Symbol: Sprr2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011474.3 | NP_035604.1 | 2 | 395 | ||
| GenBank mRNA | AJ005566.1 | - | 1 | 367 | ||
| AY158992.1 | - | 2 | 395 | |||
| BC099888.1 | - | 3 | 164 | |||
| BC099889.1 | - | 2 | 381 | |||
| BC099890.1 | - | 2 | 98 | |||
| BC127024.1 | - | 2 | 378 | |||

Sprr2h
Genomic Map