siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011485.4 | NP_035615.2 | 1, 2 | 128 | ||
| GenBank mRNA | AK014431.1 | - | 1, 2 | 138 | ||
| AK019725.2 | - | 1, 2 | 168 | |||
| AK054470.1 | - | 1, 2 | 128 | |||
| AK136327.1 | - | 1, 2 | 138 | |||
| BC060970.1 | - | 1, 2 | 139 | |||
| BC082283.1 | - | 1, 2 | 139 | 30436512 | ||
| L36062.1 | - | 1, 2 | 144 | |||

Star
Genomic Map