siRNA Details
| Gene Symbol: Stat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011486.4 | NP_035616.1 | 4 | 565 | ||
| NM_213659.2 | NP_998824.1 | 4 | 565 | |||
| NM_213660.2 | NP_998825.1 | 4 | 565 | |||
| GenBank mRNA | AK145486.1 | - | 4 | 421 | ||
| AK153005.1 | - | 4 | 477 | |||
| AK153472.1 | - | 4 | 477 | |||
| AY299489.1 | - | 3 | 292 | |||
| AY299490.1 | - | 3 | 292 | |||
| BC003806.1 | - | 4 | 453 | 3593588 | ||
| BC019168.1 | - | 4 | 469 | 4923137 | ||
| L29278.1 | - | 4 | 544 | |||
| U06922.1 | - | 4 | 354 | |||
| U08378.1 | - | 4 | 432 | |||
| U30709.1 | - | 4 | 458 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012747, XM_006247258, XM_006247257, XM_006247259, X91810, and others.

Stat3
Genomic Map