siRNA Details
| Gene Symbol: Stat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037598.1 | - | 2, 3 | 257 | ||
| AK132675.1 | - | 4, 5 | 444 | |||
| AK157083.1 | - | 3, 4 | 318 | |||
| BC052907.1 | - | 3, 4 | 323 | |||
| BC098499.1 | - | 3, 4 | 311 | 30074608 | ||
| BY750146.1 | - | 3, 4 | 309 | |||
| U06923.1 | - | 3, 4 | 314 | |||
| U09351.1 | - | 3, 4 | 768 | |||

Stat4
Genomic Map