siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011485.4 | NP_035615.2 | 3 | 316 | ||
| GenBank mRNA | AK014431.1 | - | 3 | 325 | ||
| AK019725.2 | - | 3 | 356 | |||
| AK054470.1 | - | 3 | 316 | |||
| AK136327.1 | - | 3 | 326 | |||
| BC082283.1 | - | 3 | 327 | 30436512 | ||
| L36062.1 | - | 3 | 332 | |||

Star
Genomic Map