siRNA Details
| Gene Symbol: Stk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB015801.1 | - | 4 | 557 | ||
| AF129870.1 | - | 4 | 935 | |||
| AF145287.1 | - | 4 | 539 | |||
| AF151711.1 | - | 4 | 539 | |||
| AK004499.1 | - | 2 | 218 | |||
| AK147802.1 | - | 4 | 598 | |||
| AK171909.1 | - | 5 | 1333 | |||
| AK172385.1 | - | 4 | 1322 | |||
| AK172528.1 | - | 4 | 1319 | |||
| BC052379.1 | - | 4 | 600 | 5688695 | ||
| EU730638.1 | - | 4 | 518 | |||

Stk11
Genomic Map