siRNA Details
| Gene Symbol: CHRNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000748.2 | NP_000739.1 | 3, 4 | 527 | ||
| XM_017000180.1 | XP_016855669.1 | 3, 4 | 509 | |||
| GenBank mRNA | AK313470.1 | - | 3, 4 | 296 | ||
| BC075040.2 | - | 3, 4 | 292 | 30915357 | ||
| BC075041.2 | - | 3, 4 | 292 | 30915573 | ||
| U62437.1 | - | 3, 4 | 527 | |||
| X53179.1 | - | 3, 4 | 429 | |||
| Y08415.1 | - | 3, 4 | 263 | |||

CHRNB2
Genomic Map