siRNA Details
| Gene Symbol: Spr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011467.2 | NP_035597.2 | 3 | 1044 | ||
| GenBank mRNA | AK154720.1 | - | 1048 | |||
| BC010400.1 | - | 3 | 1042 | 4219245 | ||
| BC054423.1 | - | 3 | 1028 | 4985039 | ||
| BY590544.1 | - | 1 | 287 | |||
| S77493.1 | - | 3 | 1046 | |||

Spr
Genomic Map