siRNA Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011485.4 | NP_035615.2 | 7 | 1124 | ||
| GenBank mRNA | AK014431.1 | - | 7 | 1133 | ||
| AK019725.2 | - | 7 | 1164 | |||
| AK054470.1 | - | 7 | 1124 | |||
| BC082283.1 | - | 7 | 1135 | 30436512 | ||
| L36062.1 | - | 7 | 1140 | |||

Star
Genomic Map