siRNA Details
| Gene Symbol: Tbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011536.2 | NP_035666.2 | 2 | 245 | ||
| GenBank mRNA | AB221665.1 | - | 2 | 245 | ||
| AY075134.1 | - | 2 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbx4| Gene Symbol: Tbx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011536.2 | NP_035666.2 | 2 | 245 | ||
| GenBank mRNA | AB221665.1 | - | 2 | 245 | ||
| AY075134.1 | - | 2 | 245 | |||