siRNA Details
| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011540.2 | NP_035670.2 | 2 | 170 | ||
| GenBank mRNA | AJ223854.1 | - | 2 | 170 | ||
| AK010167.1 | - | 2 | 158 | |||
| BC027631.1 | - | 2 | 142 | 1314541 | ||
| BC032992.1 | - | 2 | 142 | |||
| Y15845.1 | - | 2 | 160 | |||

Tcap
Genomic Map