siRNA Details
| Gene Symbol: Tcof1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198984.1 | NP_001185913.1 | 3 | 312 | ||
| NM_011552.3 | NP_035682.1 | 3 | 312 | |||
| GenBank mRNA | AF001794.1 | - | 3 | 256 | ||
| AK089518.1 | - | 3 | 278 | |||
| BC021514.1 | - | 3 | 317 | 5351783 | ||
| BC046608.1 | - | 3 | 266 | |||
| BC052669.1 | - | 3 | 284 | |||
| BC057342.1 | - | 3 | 265 | 6825735 | ||
| BC060105.1 | - | 3 | 265 | |||
| U81030.1 | - | 3 | 254 | |||

Tcof1
Genomic Map