siRNA Details
| Gene Symbol: Tnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011607.3 | NP_035737.2 | 2 | 664 | ||
| GenBank mRNA | AK085667.1 | - | 2 | 582 | ||
| BC117979.1 | - | 1 | 388 | 40099589 | ||
| BC117980.1 | - | 1 | 388 | 40099596 | ||
| D90343.1 | - | 2 | 465 | |||
| X56304.1 | - | 1 | 398 | |||
| X80281.1 | - | 1 | 137 | |||

Tnc
Genomic Map