siRNA Details
| Gene Symbol: Tank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164071.1 | NP_001157543.1 | 3 | 428 | ||
| NM_001164072.1 | NP_001157544.1 | 2 | 223 | |||
| NM_011529.2 | NP_035659.1 | 2 | 219 | |||
| GenBank mRNA | AK038910.1 | - | 2 | 158 | ||
| AK088817.1 | - | 2 | 169 | |||
| AK147727.1 | - | 3 | 374 | |||
| AK156658.1 | - | 1 | 46 | |||
| AK165420.1 | - | 2 | 179 | |||
| AK165994.1 | - | 2 | 154 | |||
| BB660164.1 | - | 2 | 224 | |||
| BC026148.1 | - | 2 | 204 | 3492937 | ||
| BC033282.1 | - | 2 | 155 | |||
| U51907.1 | - | 2 | 161 | |||
| U59864.1 | - | 2 | 232 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_145788, NM_001164073, XM_006234273, XM_006234274, BC129086, and others.

Tank
Genomic Map