siRNA Details
| Gene Symbol: Tbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285472.1 | NP_001272401.1 | 3, 4 | 506 | ||
| NM_011532.2 | NP_035662.1 | 1, 2 | 434 | |||
| GenBank mRNA | AF326960.2 | - | 1, 2 | 434 | ||
| AF349658.1 | - | 3, 4 | 506 | |||
| BC139474.1 | - | 1, 2 | 434 | 8862496 | ||
| U57327.1 | - | 1, 2 | 56 | |||

Tbx1
Genomic Map