siRNA Details
| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010508.2 | NP_034638.2 | 11 | 4425 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_011601.1 | - | 503 | Chr. - - [-] on Build | ||

Ifnar1
Genomic Map