siRNA Details
| Gene Symbol: Tnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011607.3 | NP_035737.2 | 3 | 1200 | ||
| GenBank mRNA | AK085667.1 | - | 3 | 1118 | ||
| BC117979.1 | - | 2 | 924 | 40099589 | ||
| BC117980.1 | - | 2 | 924 | 40099596 | ||
| D90343.1 | - | 3 | 1001 | |||
| X56304.1 | - | 2 | 934 | |||

Tnc
Genomic Map