siRNA Details
| Gene Symbol: Cd70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011617.2 | NP_035747.1 | 3 | 784 | ||
| GenBank mRNA | U78091.1 | - | 3 | 666 | ||
| Y13636.1 | - | 3 | 784 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd70| Gene Symbol: Cd70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011617.2 | NP_035747.1 | 3 | 784 | ||
| GenBank mRNA | U78091.1 | - | 3 | 666 | ||
| Y13636.1 | - | 3 | 784 | |||