siRNA Details
| Gene Symbol: Tom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136259.1 | NP_001129731.1 | 4 | 446 | ||
| NM_011622.3 | NP_035752.1 | 4 | 446 | |||
| GenBank mRNA | AJ006972.1 | - | 4 | 423 | ||
| AK028398.1 | - | 4 | 387 | |||
| AK051245.1 | - | 4 | 428 | |||
| AK150143.1 | - | 4 | 396 | |||
| AK160403.1 | - | 4 | 392 | |||
| BC021633.1 | - | 4 | 409 | 4208226 | ||
| BC093520.1 | - | 4 | 374 | 2866013 | ||
| BC157905.1 | - | 4 | 407 | 9088106 | ||
| BC158042.1 | - | 4 | 407 | 9007343 | ||

Tom1
Genomic Map