siRNA Details
Gene Symbol: Top2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011623.2 | NP_035753.2 | 4 | 421 | ||
GenBank mRNA | AK028218.1 | - | 1 | 469 | ||
BC008170.1 | - | 4 | 409 | |||
BC057873.1 | - | 4 | 391 | |||
D12513.1 | - | 4 | 347 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_022183, Z19552.
Panther Classification:
Gene Ontology Categories:
