siRNA Details
| Gene Symbol: Tesk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011571.3 | NP_035701.3 | 10 | 3394 | ||
| GenBank mRNA | AB003494.1 | - | 10 | 3732 | ||
| AK144302.1 | - | 10 | 2977 | |||
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BE692309.1 | - | 1 | 130 | ||

Tesk1
Genomic Map