siRNA Details
| Gene Symbol: Tgtp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145164.1 | NP_001138636.1 | 2 | 2682 | ||
| Gene Symbol: Tgtp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011579.3 | NP_035709.3 | 2 | 2675 | ||
| GenBank mRNA | AK089836.1 | - | 2 | 2677 | ||

Tgtp2
Genomic Map