siRNA Details
| Gene Symbol: Tia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164078.1 | NP_001157550.1 | 9, 10 | 1007 | ||
| NM_011585.4 | NP_035715.1 | 10, 11 | 1034 | |||
| GenBank mRNA | AK028340.1 | - | 8, 9 | 674 | ||
| AK088486.1 | - | 10, 11 | 970 | |||
| AK144222.1 | - | 10, 11 | 987 | |||
| AK190403.1 | - | 1, 2 | 92 | |||
| BC027826.1 | - | 8, 9 | 2971 | |||
| BC046812.1 | - | 9, 10 | 890 | 5702183 | ||
| U00689.1 | - | 10, 11 | 975 | |||
| U55862.1 | - | 10, 11 | 842 | |||

Tia1
Genomic Map