siRNA Details
| Gene Symbol: Clec3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011606.2 | NP_035736.2 | 3 | 334 | ||
| GenBank mRNA | BC035043.1 | - | 3 | 286 | 1313548 | |
| X79199.1 | - | 3 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec3b| Gene Symbol: Clec3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011606.2 | NP_035736.2 | 3 | 334 | ||
| GenBank mRNA | BC035043.1 | - | 3 | 286 | 1313548 | |
| X79199.1 | - | 3 | 309 | |||