siRNA Details
| Gene Symbol: Tnnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163664.1 | NP_001157136.1 | 12 | 393 | ||
| NM_001163665.1 | NP_001157137.1 | 12 | 393 | |||
| NM_001163666.1 | NP_001157138.1 | 10 | 366 | |||
| NM_001163667.1 | NP_001157139.1 | 11 | 381 | |||
| NM_001163668.1 | NP_001157140.1 | 10 | 342 | |||
| NM_001163669.1 | NP_001157141.1 | 11 | 354 | |||
| NM_001163670.1 | NP_001157142.1 | 8 | 309 | |||
| NM_011620.3 | NP_035750.2 | 9 | 321 | |||
| GenBank mRNA | AA028290.1 | - | 11 | 334 | ||
| AA771596.1 | - | 10 | 319 | |||
| AI173874.1 | - | 10 | 358 | |||
| AI574395.1 | - | 12 | 387 | |||
| AI746973.1 | - | 10 | 361 | |||
| AK003147.1 | - | 8 | 276 | |||
| BC003747.1 | - | 12 | 389 | 3594256 | ||
| BY766323.1 | - | 11 | 371 | |||
| CB196352.1 | - | 2 | 145 | |||
| CK791812.1 | - | 1 | 31 | |||
| DV049834.1 | - | 9 | 368 | |||
| L48988.1 | - | 6 | 212 | |||
| L48989.1 | - | 6 | 212 | |||
| L48990.1 | - | 9 | 257 | |||
| L48991.1 | - | 6 | 239 | |||
| L48992.1 | - | 8 | 245 | |||
| L48993.1 | - | 8 | 245 | |||
| L49018.1 | - | 6 | 239 | |||
| L49466.1 | - | 11 | 381 | |||
| L49467.1 | - | 12 | 393 | |||
| L49468.1 | - | 10 | 366 | |||
| L49469.1 | - | 10 | 366 | |||
| L49470.1 | - | 11 | 354 | |||
| L49471.1 | - | 10 | 363 | |||
| L49472.1 | - | 10 | 342 | |||
| U77779.1 | - | 9 | 314 | |||

Tnnt3
Genomic Map