siRNA Details
| Gene Symbol: Tmem165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011626.2 | NP_035756.2 | 3, 4 | 874 | ||
| GenBank mRNA | AK004829.1 | - | 3, 4 | 820 | ||
| AK178312.1 | - | 2, 3 | 217 | |||
| AK199397.1 | - | 2, 3 | 216 | |||
| BC037638.1 | - | 4, 5 | 655 | |||
| BC141081.1 | - | 3, 4 | 658 | 9055693 | ||
| M23568.1 | - | 3, 4 | 805 | |||

Tmem165
Genomic Map