siRNA Details
| Gene Symbol: Tulp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011657.2 | NP_035787.1 | 3 | 257 | ||
| GenBank mRNA | AB016963.1 | - | 3 | 200 | ||
| AF045582.1 | - | 3 | 210 | |||
| AK035363.1 | - | 3 | 222 | |||
| AK142527.1 | - | 3 | 247 | |||
| BC060068.1 | - | 3 | 187 | 6419643 | ||
| BY347356.1 | - | 3 | 260 | |||

Tulp3
Genomic Map