siRNA Details
| Gene Symbol: Twist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011658.2 | NP_035788.1 | 1 | 788 | ||
| GenBank mRNA | BC033434.1 | - | 1 | 614 | 4935230 | |
| BC083139.1 | - | 1 | 788 | 6516673 | ||
| M63650.1 | - | 1 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Twist1| Gene Symbol: Twist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011658.2 | NP_035788.1 | 1 | 788 | ||
| GenBank mRNA | BC033434.1 | - | 1 | 614 | 4935230 | |
| BC083139.1 | - | 1 | 788 | 6516673 | ||
| M63650.1 | - | 1 | 647 | |||