siRNA Details
| Gene Symbol: Usp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011678.2 | NP_035808.2 | 3 | 347 | ||
| GenBank mRNA | AK138838.1 | - | 3 | 282 | ||
| AK143582.1 | - | 3 | 282 | |||
| AK149964.1 | - | 3 | 305 | |||
| AK169933.1 | - | 3 | 317 | |||
| AK171271.1 | - | 3 | 317 | |||
| AK171633.1 | - | 3 | 300 | |||
| AK214431.1 | - | 2 | 143 | |||
| AK218565.1 | - | 2 | 143 | |||
| AK220512.1 | - | 3 | 304 | |||
| BC011341.1 | - | 3 | 265 | 3661165 | ||
| BC066180.1 | - | 3 | 257 | |||
| BC066865.1 | - | 3 | 257 | |||
| L00681.1 | - | 3 | 310 | |||

Usp4
Genomic Map