siRNA Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185164.1 | NP_001172093.1 | 5 | 635 | ||
| NM_011697.3 | NP_035827.1 | 5 | 635 | |||
| GenBank mRNA | AK148188.1 | - | 5 | 634 | ||
| AK182808.1 | - | 3 | 263 | |||
| AK207652.1 | - | 3 | 263 | |||
| AK208173.1 | - | 3 | 266 | |||
| AK211743.1 | - | 3 | 267 | |||
| AK212120.1 | - | 3 | 266 | |||
| BC046303.1 | - | 5 | 613 | 6492277 | ||
| U43836.1 | - | 5 | 587 | |||
| U43837.1 | - | 5 | 395 | |||
| U48800.1 | - | 5 | 489 | |||
| U52820.1 | - | 5 | 489 | |||

Vegfb
Genomic Map