siRNA Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317384.1 | NP_001304313.1 | 2 | 486 | ||
| NM_001317387.1 | NP_001304316.1 | 2 | 486 | |||
| NM_001650.5 | NP_001641.1 | 2 | 486 | |||
| NM_004028.4 | NP_004019.1 | 1 | 613 | |||
| XM_011525942.2 | XP_011524244.1 | 2 | 528 | |||
| GenBank mRNA | AB209156.1 | - | 2 | 352 | ||
| AK222684.1 | - | 2 | 398 | |||
| AK295069.1 | - | 3 | 297 | |||
| BC022286.1 | - | 2 | 351 | 4717755 | ||
| BI596912.1 | - | 2 | 386 | |||
| BI667387.1 | - | 2 | 404 | |||
| D63412.1 | - | 2 | 351 | |||
| HQ901095.1 | - | 2 | 460 | |||
| KF055862.1 | - | 2 | 331 | |||
| U34845.1 | - | 1 | 549 | |||
| U63622.1 | - | 2 | 370 | |||
| U63623.1 | - | 1 | 274 | |||

AQP4
Genomic Map